ENA311759779 Enamine

2-(5-fluoro-2-methoxyphenyl)-2-oxoacetic acid

  • MDL number MFCD11179658
  • Cas Number : 1094355-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71308
Availability details for
Quantity
  • Availability
  • Add to Cart