ENA311759781 Enamine

2-[(5-bromopyridin-2-yl)amino]-2-methylpropan-1-ol

  • MDL number MFCD12184825
  • Cas Number : 1154894-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71311
Availability details for
Quantity
  • Availability
  • Add to Cart