ENA311759783 Enamine

2-(2,3-dichlorophenoxy)-2-phenylacetic acid

  • MDL number MFCD09704702
  • Cas Number : 938116-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71313

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71313
Availability details for
Quantity
  • Availability
  • Add to Cart