ENA311759786 Enamine

quinoline-5-carbohydrazide

  • MDL number MFCD01063087
  • Cas Number : 96541-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71316

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71316
Availability details for
Quantity
  • Availability
  • Add to Cart