ENA311759788 Enamine

5-hydroxypyridine-2-carbaldehyde

  • MDL number MFCD10697538
  • Cas Number : 31191-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-184
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71319
Availability details for
Quantity
  • Availability
  • Add to Cart