ENA311759791 Enamine

5-fluoro-2-(1H-pyrazol-1-yl)aniline

  • MDL number MFCD08699509
  • Cas Number : 1006960-35-5
  • Molecular Weight : 177.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71322
Availability details for
Quantity
  • Availability
  • Add to Cart