ENA311759808 Enamine

2-amino-1-(2-methoxyphenyl)ethan-1-one hydrochloride

  • MDL number MFCD11974304
  • Cas Number : 34589-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71348

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71348
Availability details for
Quantity
  • Availability
  • Add to Cart