ENA311759812 Enamine

methyl 3-ethynylpyridine-2-carboxylate

  • MDL number MFCD13189769
  • Cas Number : 1196156-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71369

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71369
Availability details for
Quantity
  • Availability
  • Add to Cart