ENA311759819 Enamine

N-(4-cyanophenyl)-2-cyclopentylacetamide

  • MDL number MFCD09951681
  • Cas Number : 1016862-67-1
  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71384
Availability details for
Quantity
  • Availability
  • Add to Cart