ENA311759824 Enamine

1-methanesulfonyl-3-(prop-2-yn-1-yloxy)benzene

  • MDL number MFCD14631686
  • Cas Number : 1250110-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71390
Availability details for
Quantity
  • Availability
  • Add to Cart