ENA311759828 Enamine

1-(2-benzyl-4-methyl-1,3-thiazol-5-yl)ethan-1-one

  • MDL number MFCD08554746
  • Cas Number : 937598-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53...55
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71394

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71394
Availability details for
Quantity
  • Availability
  • Add to Cart