ENA311759829 Enamine

1-[2-(4-chlorophenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-amine

  • MDL number MFCD09882222
  • Cas Number : 1017170-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-71395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71395
Availability details for
Quantity
  • Availability
  • Add to Cart