ENA311759832 Enamine

4-chloro-2-(propan-2-yl)pyrimidine

  • MDL number MFCD09746255
  • Cas Number : 68210-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71398

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-71398
pictograms GHS07
hstatements H302,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart