ENA311759833 Enamine

1-(2-hydroxyethyl)piperidin-2-one

  • MDL number MFCD12186793

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71399

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71399
Availability details for
Quantity
  • Availability
  • Add to Cart