ENA311759839 Enamine

1-(2,4-difluorophenyl)pyrrolidin-3-amine

  • MDL number MFCD11640823
  • Cas Number : 1096333-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71408
Availability details for
Quantity
  • Availability
  • Add to Cart