ENA311759842 Enamine

2-chloro-N-(cyclohexylmethyl)-N-methylacetamide

  • MDL number MFCD12762660
  • Cas Number : 895244-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-46846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46846
Availability details for
Quantity
  • Availability
  • Add to Cart