ENA311759850 Enamine

2-[(3-acetylbenzene)sulfonamido]-3-carbamoylpropanoic acid

  • MDL number MFCD13352672

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191...193
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71423

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71423
Availability details for
Quantity
  • Availability
  • Add to Cart