ENA311759857 Enamine

ethyl 2-(3-chloropyridin-2-yl)-5-oxopyrazolidine-3-carboxylate

  • MDL number MFCD09955582
  • Cas Number : 500011-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71438
Availability details for
Quantity
  • Availability
  • Add to Cart