ENA311759863 Enamine

4-bromo-3-(4-fluorophenyl)-1H-pyrazole

  • MDL number MFCD12032231
  • Cas Number : 474706-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-68741

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68741
Availability details for
Quantity
  • Availability
  • Add to Cart