ENA311759865 Enamine

2-(4-chlorophenyl)-2-(dimethylamino)propanoic acid

  • MDL number MFCD05230699
  • Cas Number : 1157479-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68744
Availability details for
Quantity
  • Availability
  • Add to Cart