ENA311759867 Enamine

1-[2-(2-bromophenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-one

  • MDL number MFCD14615022
  • Cas Number : 1248601-42-4
  • Molecular Weight : 296.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68817
Availability details for
Quantity
  • Availability
  • Add to Cart