ENA311759871 Enamine

3-(dipropylcarbamoyl)bicyclo[2.2.1]hept-5-ene-2-carboxylic acid

  • MDL number MFCD00985432
  • Cas Number : 1005121-21-0
  • Molecular Weight : 265.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71458
Availability details for
Quantity
  • Availability
  • Add to Cart