ENA311759882 Enamine

1-(4-chlorophenyl)-5-phenylpentan-2-one

  • MDL number MFCD16350622
  • Cas Number : 1179919-66-4
  • Molecular Weight : 272.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68954
Availability details for
Quantity
  • Availability
  • Add to Cart