ENA311759883 Enamine

(4-fluorophenyl)(pyridin-4-yl)methanamine

  • MDL number MFCD09026464
  • Cas Number : 917746-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68958
Availability details for
Quantity
  • Availability
  • Add to Cart