ENA311759884 Enamine

1-[(oxolan-2-yl)carbonyl]piperidine-2-carboxylic acid

  • MDL number MFCD08444914
  • Cas Number : 1032057-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71472

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71472
Availability details for
Quantity
  • Availability
  • Add to Cart