ENA311759892 Enamine

1-(4-amino-1H-pyrazol-1-yl)-3-(pyrrolidin-1-yl)propan-2-ol

  • MDL number MFCD11654971
  • Cas Number : 1152896-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71480
Availability details for
Quantity
  • Availability
  • Add to Cart