ENA311759895 Enamine

4-(benzyloxy)-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD18089392
  • Cas Number : 86045-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71483

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71483
Availability details for
Quantity
  • Availability
  • Add to Cart