ENA311759902 Enamine

5,7-dimethoxy-4-oxo-1,4-dihydroquinoline-3-carbonitrile

  • MDL number MFCD09951496
  • Cas Number : 331662-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71492
Availability details for
Quantity
  • Availability
  • Add to Cart