ENA311759908 Enamine

1-(3-chlorophenyl)-2,2,2-trifluoroethan-1-ol

  • MDL number MFCD00461901
  • Cas Number : 81577-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-71498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71498
Availability details for
Quantity
  • Availability
  • Add to Cart