ENA311759922 Enamine

butyl 2-aminoacetate hydrochloride

  • MDL number MFCD00151824
  • Cas Number : 13048-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71524
Availability details for
Quantity
  • Availability
  • Add to Cart