ENA311759923 Enamine

N-cyclohexyl-4-ethyl-1,2,3-thiadiazole-5-carboxamide

  • MDL number MFCD10561506
  • Cas Number : 1017334-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-71526

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71526
Availability details for
Quantity
  • Availability
  • Add to Cart