ENA311759929 Enamine

6-methyl-3,4-dihydro-2H-1-benzothiopyran-4-amine

  • MDL number MFCD11207727
  • Cas Number : 1094350-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 85
Product Link https://www.enaminestore.com/catalog/EN300-71535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71535
Availability details for
Quantity
  • Availability
  • Add to Cart