ENA311759932 Enamine

4-chloro-1-(2,2-diethoxyethyl)-1H-pyrazole

  • MDL number MFCD08445404
  • Cas Number : 1006334-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71538

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71538
Availability details for
Quantity
  • Availability
  • Add to Cart