ENA311760277 Enamine

3,5-dimethyl-1H-pyrazole-4-carboxylic acid hydrochloride

  • MDL number MFCD17977152
  • Cas Number : 1290998-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71562

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71562
Availability details for
Quantity
  • Availability
  • Add to Cart