ENA311760278 Enamine

propyl 3-aminopropanoate hydrochloride

  • MDL number MFCD17977153
  • Cas Number : 81440-29-1
  • Molecular Weight : 167.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71579
Availability details for
Quantity
  • Availability
  • Add to Cart