ENA311760279 Enamine

3-{[1-(4-bromophenyl)-1-oxopropan-2-yl]carbamoyl}propanoic acid

  • MDL number MFCD18089396
  • Cas Number : 1305712-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-71581

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71581
Availability details for
Quantity
  • Availability
  • Add to Cart