ENA311760281 Enamine

4-(4-methoxyphenyl)-1,2,3,6-tetrahydropyridine hydrochloride

  • MDL number MFCD18089399
  • Cas Number : 143017-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71608
Availability details for
Quantity
  • Availability
  • Add to Cart