ENA311760287 Enamine

3-methyl-1H-pyrazole-4-carbohydrazide

  • MDL number MFCD18089403
  • Cas Number : 1308650-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71653
Availability details for
Quantity
  • Availability
  • Add to Cart