ENA311760290 Enamine

2-{3-[(dimethylamino)methyl]phenyl}ethan-1-amine dihydrochloride

  • MDL number MFCD18089409
  • Cas Number : 1305712-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71698
Availability details for
Quantity
  • Availability
  • Add to Cart