ENA311760306 Enamine

3-{3H-imidazo[4,5-b]pyridin-2-yl}propanoic acid dihydrochloride

  • MDL number MFCD18089425
  • Cas Number : 1303890-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71876

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-71876
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart