ENA311760312 Enamine

benzyl 3-[(chlorosulfonyl)methyl]piperidine-1-carboxylate

  • MDL number MFCD18089431
  • Cas Number : 242459-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81...83
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71924

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71924
Availability details for
Quantity
  • Availability
  • Add to Cart