ENA311760314 Enamine

2-amino-N-(2-hydroxyethyl)benzamide hydrochloride

  • MDL number MFCD18089432
  • Cas Number : 34637-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71948
Availability details for
Quantity
  • Availability
  • Add to Cart