ENA311760322 Enamine

butyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-carbamoylbutanoate

  • MDL number MFCD18089441
  • Cas Number : 1308958-08-8
  • Molecular Weight : 302.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72020
Availability details for
Quantity
  • Availability
  • Add to Cart