ENA311760326 Enamine

methyl 3-amino-2-[(1-methyl-1H-pyrazol-4-yl)methyl]propanoate dihydrochloride

  • MDL number MFCD18089448
  • Cas Number : 1305712-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72050

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72050
Availability details for
Quantity
  • Availability
  • Add to Cart