ENA311760334 Enamine

methyl 3-amino-2-[(pyridin-4-yl)methyl]propanoate dihydrochloride

  • MDL number MFCD18089458
  • Cas Number : 1303889-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72127
Availability details for
Quantity
  • Availability
  • Add to Cart