ENA311760336 Enamine

1-(difluoromethyl)-3,5-dimethyl-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD18089461
  • Cas Number : 1305711-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72172

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72172
Availability details for
Quantity
  • Availability
  • Add to Cart