ENA311760345 Enamine

2-cyclopentyl-1-(1,4-diazepan-1-yl)ethan-1-one hydrochloride

  • MDL number MFCD18089471
  • Cas Number : 1148033-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72205

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72205
Availability details for
Quantity
  • Availability
  • Add to Cart