ENA311760349 Enamine

N-amino-3,5-difluorobenzene-1-carboximidamide

  • MDL number MFCD18089476
  • Cas Number : 1251448-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72240
Availability details for
Quantity
  • Availability
  • Add to Cart