ENA311760352 Enamine

2-chloro-1-{4-[(furan-2-yl)methyl]piperazin-1-yl}ethan-1-one hydrochloride

  • MDL number MFCD18089479
  • Cas Number : 1303889-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72254
Availability details for
Quantity
  • Availability
  • Add to Cart