ENA311760366 Enamine

2-chloro-1-{4-[(thiophen-3-yl)methyl]piperazin-1-yl}ethan-1-one hydrochloride

  • MDL number MFCD18089494
  • Cas Number : 1308650-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72371
Availability details for
Quantity
  • Availability
  • Add to Cart